Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563814
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563814 Supplier Invitrogen™ Supplier No. PA563814
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NTKMRSSNSA SASAGPPGKA QTQSPHLHPH PHPSTSTPVP SSI Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HOXA4
Gene Accession No. Q00056
Gene Alias AV206827; Dfd-like protein; homeo box A4; homeobox A4; homeobox protein Hox-1.4; Homeobox protein Hox-1D; homeobox protein Hox-A4; homeobox protein Hox-A4-like; homeobox protein MH-3; HOX1; Hox-1.4; Hox-1.4-like protein; Hox-14; HOX1D; Hoxa4; Hoxa-4; LOC100912525
Gene Symbols HOXA4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HOXA4. Recombinant protein control fragment (Product #RP-100085).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3201
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.