Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXA5 Antibody (SR1735), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | HOXA5 |
---|---|
Clone | SR1735 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
HOXA5 Monoclonal antibody specifically detects HOXA5 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
HOXA5 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
homeo box 1C, homeo box A5, homeobox A5, Homeobox protein Hox-1C, homeobox protein Hox-A5, homeobox protein HOXA5, HOX1, HOX1.3, HOX1CMGC9376 | |
A synthesized peptide derived from human HOXA5 (Uniprot #: P20719) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1735 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3202 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title