Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXA5 Monoclonal Antibody (1D12)
GREENER_CHOICE

Catalog No. PIMA519103
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA519103 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA519103 Supplier Invitrogen™ Supplier No. MA519103
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: PAQPQIYPWM RKLHISHDNI GGPEGKRART AYTRYQTLEL EKEFHFNRYL TRRRRIEIAH ALCLSERQIK IWFQNRRMKW KKDNKLKSMS MAAAGGAFRP.

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. HOXA5 gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. Methylation of this gene may result in the loss of its expression and, since the encoded protein upregulates the tumor suppressor p53, this protein may play an important role in tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA5
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1D12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene HOXA5
Gene Accession No. P20719
Gene Alias homeo box 1C; homeo box A5; homeobox A5; homeobox protein Hox-1.3; Homeobox protein Hox-1C; homeobox protein HOXA5; homeobox protein Hox-A5; homeobox protein M2; HOX1; HOX1.3; Hox-1.3; HOX1C; Hox-1C; Hoxa5; Hoxa-5; Unknown (protein for MGC:138007)
Gene Symbols HOXA5
Host Species Mouse
Immunogen HOXA5 (NP_061975.1, 171 a.a. ~ 270 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3202
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.