Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583081 Supplier Invitrogen™ Supplier No. PA583081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MHSGRYGYGY NGMDLSVGRS GSGHFGSGER ARSYAASASA APAEPRYSQP ATSTHSPQPD PLPCSAVAPS PGSDSHHGGK NSLSNSSGAS ADAGSTHISS REGVGTASGA EEDAPASSEQ ASAQSEPSPA PPAQPQIYPW MRKLHISHDN Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 96%.

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. HOXA5 gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. Methylation of this gene may result in the loss of its expression and, since the encoded protein upregulates the tumor suppressor p53, this protein may play an important role in tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HOXA5
Gene Accession No. P20719
Gene Alias homeo box 1C; homeo box A5; homeobox A5; homeobox protein Hox-1.3; Homeobox protein Hox-1C; homeobox protein HOXA5; homeobox protein Hox-A5; homeobox protein M2; HOX1; HOX1.3; Hox-1.3; HOX1C; Hox-1C; Hoxa5; Hoxa-5; Unknown (protein for MGC:138007)
Gene Symbols HOXA5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HOXA5. Recombinant protein control fragment (Product #RP-102492).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3202
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.