Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXA5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569008
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. HOXA5 gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. Methylation of this gene may result in the loss of its expression and, since the encoded protein upregulates the tumor suppressor p53, this protein may play an important role in tumorigenesis.
Specifications
HOXA5 | |
Polyclonal | |
Unconjugated | |
HOXA5 | |
homeo box 1C; homeo box A5; homeobox A5; homeobox protein Hox-1.3; Homeobox protein Hox-1C; homeobox protein HOXA5; homeobox protein Hox-A5; homeobox protein M2; HOX1; HOX1.3; Hox-1.3; HOX1C; Hox-1C; Hoxa5; Hoxa-5; Unknown (protein for MGC:138007) | |
Rabbit | |
Affinity chromatography | |
RUO | |
15402, 3202 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Surface Plasmon Resonance | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P09021, P20719 | |
HOXA5 | |
synthetic peptide directed towards the C terminal region of human HOXA5. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction