Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXA5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986741
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6741 Supplier Bioss Supplier No. BS5713R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. HOXA5 gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. Methylation of this gene may result in the loss of its expression and, since the encoded protein upregulates the tumor suppressor p53, this protein may play an important role in tumorigenesis.

Specifications

Antigen HOXA5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HOXA5
Gene Accession No. P20719
Gene Alias BOS_4327; homeo box 1C; homeo box A5; homeobox A5; Homeobox protein Hox-1.3; Homeobox protein Hox-1C; homeobox protein HOXA5; Homeobox protein Hox-A5; Homeobox protein M2; HOX1; HOX1.3; Hox-1.3; HOX1C; Hox-1C; HOXA5; Hoxa-5
Gene Symbols HOXA5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HOXA5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3202
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.