Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXA6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579390
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579390 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579390 Supplier Invitrogen™ Supplier No. PA579390
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: SW620 cell, SW620 cell, HELA cell, HT1080 cell, HT1080 cell.

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA6
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene HOXA6
Gene Accession No. P09092, P31267
Gene Alias homeo box 1B; homeo box A6; homeobox A6; homeobox protein Hox-1.2; Homeobox protein Hox-1B; homeobox protein HOXA6; homeobox protein Hox-A6; homeobox protein M5-4; HOX1; HOX1.2; Hox-1.2; HOX1B; Hoxa6; Hoxa-6
Gene Symbols HOXA6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human HOXA6 (95-114aa KQRGPGDYLHFSPEQQYKPD).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15403, 3203, 685732
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.