Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXA6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597836
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597836 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597836 Supplier Invitrogen™ Supplier No. PA597836
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOXA6 Polyclonal Antibody for Western Blot, IHC (P), ELISA

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene HOXA6
Gene Accession No. P09092, P31267
Gene Alias homeo box 1B; homeo box A6; homeobox A6; homeobox protein Hox-1.2; Homeobox protein Hox-1B; homeobox protein HOXA6; homeobox protein Hox-A6; homeobox protein M5-4; HOX1; HOX1.2; Hox-1.2; HOX1B; Hoxa6; Hoxa-6
Gene Symbols HOXA6
Host Species Rabbit
Immunogen Recombinant Human Homeobox protein Hox-A6 protein (1-233AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15403, 3203
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.