Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXA7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567324
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567324 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567324 Supplier Invitrogen™ Supplier No. PA567324
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VNALFSKYTAG TSLFQNAEPT SCSFAPNSQR SGYGA Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.

Hox genes play a fundamental role in the development of the vertebrate central nervous system, heart, axial skeleton, limbs, gut, urogenital tract and external genitalia. HoxA2 protein may contribute to dorsal-ventral patterning and/or to the specification of neuronal phenotype. HoxA7 functions as a potent transcriptional repressor and its action as such requires several domains, including both activator and repressor regions. HoxA7 is expressed in the fetal liver, lung, skeletal muscle, kidney, pancreas and placenta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA7
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HOXA7
Gene Accession No. P31268
Gene Alias ANTP; AV118143; homeo box A7; homeobox A7; homeobox A9; Homeobox gene A7; homeobox protein Hox 1.1; Homeobox protein Hox-1.1; homeobox protein HOX-1A; Homeobox protein Hox-A7; HOMEOBOX PROTEIN HOX-A7 (HOX-1.1) (M6-12) (M6); Homeobox protein M6-12; homeobox protein R5; HOX1; HOX1.1; hox-1.1; HOX1A; Hox1r5; Hoxa7; Hoxa-7; Hoxa9; M6
Gene Symbols HOXA7
Host Species Rabbit
Immunogen Recombinant Human HOXA7. Recombinant protein control fragment (Product #RP-107967).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3204
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.