Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXA9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119784
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119784 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119784 Supplier Invitrogen™ Supplier No. PA5119784
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: HepG2 cell lysate, mouse kidney tissue lysate, mouse kidney tissue. Subcellular Location: Nucleus.

HOXA9 is the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Hoxa9
Gene Accession No. P09631, P31269
Gene Alias ABD-B; D6a9; homeo box A9; homeobox A9; homeobox A9-like; homeobox protein Hox-1.7; Homeobox protein Hox-1G; homeobox protein Hox-A9; homeodomain protein HOXA9; HOX1; HOX1.7; Hox-1.7; HOX1G; HOXA 9; Hoxa9; Hoxa-9; Hoxa-9 protein; Hoxa9l; RGD1310001
Gene Symbols Hoxa9
Host Species Rabbit
Immunogen Synthetic peptide within human HOXA9 aa 20-60.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15405, 3205
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.