Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXA9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595985 Supplier Invitrogen™ Supplier No. PA595985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATTGALGNY YVDSFLLGAD AADELSVGRY APGTLGQPPR QAATLAEHPD FSPCSFQSKA TVFGASWNPV HAAGANAVPA AVYHHHHHHP YVHPQAPVAA AAPDGRYMRS WLEPTPGALS FAGLPSSRPY GIKPEPLSAR RGDCPTLDTH TLSLTDYACG SPPVDREKQP SEGAFSENNA ENESGGDKPP; Positive Samples: SW620; Cellular Location: Nucleus.

HOXA9 is the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Hoxa9
Gene Accession No. P09631, P31269
Gene Alias ABD-B; D6a9; homeo box A9; homeobox A9; homeobox A9-like; homeobox protein Hox-1.7; Homeobox protein Hox-1G; homeobox protein Hox-A9; homeodomain protein HOXA9; HOX1; HOX1.7; Hox-1.7; HOX1G; HOXA 9; Hoxa9; Hoxa-9; Hoxa-9 protein; Hoxa9l; RGD1310001
Gene Symbols Hoxa9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human HOXA9 (NP_6899521).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15405, 3205
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.