Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXA9 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501992885
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2885 100 μL
50-199-2886 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2885 Supplier Active Motif Supplier No. 39825
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOXA9 is the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development.

Specifications

Antigen HOXA9
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene Hoxa9
Gene Accession No. P31269
Gene Alias ABD-B; D6a9; homeo box A9; homeobox A9; homeobox A9-like; Homeobox protein Hox-1.7; Homeobox protein Hox-1G; Homeobox protein Hox-A9; homeodomain protein HOXA9; HOX1; HOX1.7; Hox-1.7; HOX1G; HOXA9; Hoxa-9; Hoxa9l; RGD1310001
Gene Symbols Hoxa9
Host Species Rabbit
Immunogen This HOXA9 antibody was raised against a peptide within the N-terminal region of human HOXA9.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3205
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.