Learn More
HOXB1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | HOXB1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2216
![]() ![]() |
Proteintech Group Inc
187321AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
18732-1-AP is a rabbit polyclonal antibody raised against a peptide mapping within an internal region of human HOXB1.
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis. Acts on the anterior body structures.Specifications
HOXB1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P14653, P17919 | |
15407, 3211 | |
Peptide | |
Primary | |
-20°C | |
HOXB1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
homeobox B1, Homeobox protein Hox 2I, Homeobox protein Hox B1, Hox 2.9, HOX2, HOX2I, HOXB1 | |
HOXB1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.