Learn More
Invitrogen™ HOXB13 Polyclonal Antibody

Description
Immunogen sequence: YATLDGAKDI EGLLGAGGGR NLVAHSPLTS HPAAPTLMPA VNYAPLDLPG SAEPPKQCHP CPGVPQGTSP APVPYGY Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.
Specifications
Specifications
| Antigen | HOXB13 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | HOXB13 |
| Gene Accession No. | Q92826 |
| Gene Alias | homeo box B13; homeobox B13; homeobox protein Hox-B13; HOXA13; HOXB13; PSGD |
| Gene Symbols | HOXB13 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.