Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXB13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598698
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598698 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598698 Supplier Invitrogen™ Supplier No. PA598698
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOXB13 Polyclonal Antibody for Western Blot, ELISA

HOXB13 encodes a transcription factor that belongs to the homeobox gene family. Genes of this family are highly conserved among vertebrates and essential for vertebrate embryonic development. This gene has been implicated to play a role in fetal skin development and cutaneous regeneration. In mice, a similar gene was shown to exhibit temporal and spatial colinearity in the main body axis of the embryo, but was not expressed in the secondary axes, which suggests functions in body patterning along the axis. This gene and other HOXB genes form a gene cluster at chromosome the 17q21-22 region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXB13
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene HOXB13
Gene Accession No. P70321, Q92826
Gene Alias homeo box B13; homeobox B13; homeobox protein Hox-B13; HOXA13; HOXB13; PSGD
Gene Symbols HOXB13
Host Species Rabbit
Immunogen Recombinant Human Homeobox protein Hox-B13 protein (1-161AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10481, 15408
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.