Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXB2 Mouse anti-Human, Clone: PCRP-HOXB2-1F2, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313937100UG
This item is not returnable.
View return policy
Description
HOXB2 Monoclonal antibody specifically detects HOXB2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Protein ArraySpecifications
HOXB2 | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
P14652 | |
Mouse | |
Protein A or G purified | |
RUO | |
3212 | |
Store at 4°C. | |
IgG2b |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Peptide Array | |
PCRP-HOXB2-1F2 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
homeo box 2H, homeo box B2, homeobox B2, Homeobox protein Hox-2.8, Homeobox protein Hox-2H, homeobox protein Hox-B2, HOX2, HOX2HHox-2.8, K8, K8 home protein | |
Recombinant fragment (aa114-220) of human HOXB2 (Uniprot: P14652) | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction