Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565483
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565483 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565483 Supplier Invitrogen™ Supplier No. PA565483
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QTFPSLQPGAS TLQRPRSQKR AED Highest antigen sequence identity to the following orthologs - mouse 83%, rat 83%.

Hoxb-1 maintains this function by acting very early during hindbrain neurogenesis to specify effectors of the sonic hedgehog and Mash1 signaling pathways. Hoxb2 is a homeodomain protein important in neural development that is also expressed during erythropoiesis, hindbrain development and normal human adult lung development. Hoxb2 may modulate the amount of gamma-globin mRNA expressed during development and differentiation. In addition, Hoxb2 plays an important role in the patterning of hindbrain and pharyngeal arches in the zebrafish.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXB2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HOXB2
Gene Accession No. P14652
Gene Alias AI894218; homeo box 2H; homeo box B2; homeo box B2-like; homeobox B2; homeobox protein Hox-2.8; Homeobox protein Hox-2H; homeobox protein Hox-B2; HOX2; Hox-2.8; HOX2H; Hoxb2; Hoxb2-like epididymal protein; Hoxbes2; K8; K8 home protein
Gene Symbols HOXB2
Host Species Rabbit
Immunogen Recombinant Human HOXB2. Recombinant protein control fragment (Product #RP-104839).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3212
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.