Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXB2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987320
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7320 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7320 Supplier Bioss Supplier No. BS6538R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Hoxb-1 maintains this function by acting very early during hindbrain neurogenesis to specify effectors of the sonic hedgehog and Mash1 signaling pathways. Hoxb2 is a homeodomain protein important in neural development that is also expressed during erythropoiesis, hindbrain development and normal human adult lung development. Hoxb2 may modulate the amount of gamma-globin mRNA expressed during development and differentiation. In addition, Hoxb2 plays an important role in the patterning of hindbrain and pharyngeal arches in the zebrafish.

Specifications

Antigen HOXB2
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HOXB2
Gene Accession No. P14652
Gene Alias AI894218; BOS_19013; homeo box 2H; homeo box B2; homeo box B2-like; homeobox B2; Homeobox protein Hox-2.8; Homeobox protein Hox-2H; Homeobox protein Hox-B2; HOX2; Hox-2.8; HOX2H; Hoxb2; Hoxb2-like epididymal protein; Hoxbes2; K8; K8 home protein; LOC100347179
Gene Symbols HOXB2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HOXB2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3212
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.