Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXB2 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | HOXB2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
HOXB2 Polyclonal antibody specifically detects HOXB2 in Mouse, Rat samples. It is validated for Western BlotSpecifications
HOXB2 | |
Polyclonal | |
Purified | |
RUO | |
PBS, 50% glycerol, pH7.3 | |
3212 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
homeo box 2H, homeo box B2, homeobox B2, Homeobox protein Hox-2.8, Homeobox protein Hox-2H, homeobox protein Hox-B2, HOX2, HOX2HHox-2.8, K8, K8 home protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 50-140 of human HOXB2 (NP_002136.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title