Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXB3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568211
Description
Predicted to react with Mouse and Rat samples.
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
Specifications
HOXB3 | |
Polyclonal | |
Unconjugated | |
HOXB3 | |
homeo box 2G; homeo box B3; homeobox B3; homeobox protein Hox-2.7; Homeobox protein Hox-2G; homeobox protein Hox-B3; Homeobox protein MH-23; HOX2; Hox-2.7; HOX2G; HOXB3; Hoxb-3 | |
Rabbit | |
affinity chromatography | |
RUO | |
15410, 303488, 3213 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P09026, P14651 | |
HOXB3 | |
Synthesized peptide derived from human HOXB3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction