Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXB4 Monoclonal Antibody (3A2F2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531768
Description
MA5-31768 has been tested in indirect ELISA.
HOX homeobox genes play a role both in the early stem cell function as well as in later stages of hematopoietic differentiation. HOXB4, specifically, is a homeotic selector gene implicated in self-renewal of definitive HSCs. Expression of HOXB4 in primitive progenitors combined with culture on hematopoietic stroma induced a switch to the definitive HSC phenotype. Activation of WNT signaling in HSCs induced increased expression of HOXB4. Perturbations of HOX gene expression can cause leukemia.
Specifications
HOXB4 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P17483 | |
HOXB4 | |
Purified recombinant fragment of human HOXB4 (AA: 16-251) expressed in E. coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3A2F2 | |
Unconjugated | |
HOXB4 | |
homeo box 2F; homeo box B4; homeo box B4a; homeobox B4; homeobox B4a; homeobox gene B-4; homeobox protein Hox-2.6; Homeobox protein Hox-2F; homeobox protein Hox-B4; Homeobox protein Hox-B4a; homeobox protein Zf-13; HOX2; Hox-2.6; HOX2F; Hoxb4; HOX-B4; Hoxb-4; Hoxb4 protein; hoxb4a; Hoxb4a variant 2; RGD1560113; sb:eu512; Z-17; zf13; ZF-13 | |
Mouse | |
Protein G | |
RUO | |
3214 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction