Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXB4 Recombinant Rabbit Monoclonal Antibody (6M4G5)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA537955
Description
Positive Samples: 293T, K-562 Immunogen sequence: MAMSSFLINS NYVDPKFPPC EEYSQSDYLP SDHSPGYYAG GQRRESSFQP EAGFGRRAAC TVQRYAACRD PGPPPPPPPP PPPPPPPGLS PRAPAPPPAG.
HOX homeobox genes play a role both in the early stem cell function as well as in later stages of hematopoietic differentiation. HOXB4, specifically, is a homeotic selector gene implicated in self-renewal of definitive HSCs. Expression of HOXB4 in primitive progenitors combined with culture on hematopoietic stroma induced a switch to the definitive HSC phenotype. Activation of WNT signaling in HSCs induced increased expression of HOXB4. Perturbations of HOX gene expression can cause leukemia.
Specifications
HOXB4 | |
Recombinant Monoclonal | |
1.09 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P10284, P17483 | |
HOXB4 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HOXB4 (P17483). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
6M4G5 | |
Unconjugated | |
HOXB4 | |
homeo box 2F; homeo box B4; homeo box B4a; homeobox B4; homeobox B4a; homeobox gene B-4; homeobox protein Hox-2.6; Homeobox protein Hox-2F; homeobox protein Hox-B4; Homeobox protein Hox-B4a; homeobox protein Zf-13; HOX2; Hox-2.6; HOX2F; Hoxb4; HOX-B4; Hoxb-4; Hoxb4 protein; hoxb4a; Hoxb4a variant 2; RGD1560113; sb:eu512; Z-17; zf13; ZF-13 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
15412, 3214, 497988 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction