Learn More
Invitrogen™ HOXB5 Monoclonal Antibody (3F10)

Description
Peptide Sequence: EGQTPQIFPW MRKLHISHDM TGPDGKRART AYTRYQTLEL EKEFHFNRYL TRRRRIEIAH ALCLSERQIK IWFQNRRMKW KKDNKLKSMS LATAGSAF.
Specifications
Specifications
| Antigen | HOXB5 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3F10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | HOXB5 |
| Gene Accession No. | P09067 |
| Gene Alias | AI385717; HHO.C10; homeo box 2A; homeo box B5; homeobox B5; homeobox protein H24.1; Homeobox protein HHO.C10; homeobox protein Hox-2.1; Homeobox protein Hox-2A; homeobox protein Hox-B5; Homeobox protein Hu-1; homeobox protein Mu-1; HOX2; Hox2.1; Hox-2.1; HOX2A; HOXB5; Hoxb-5; HU-1; RGD1562292 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.