Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXB5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540375
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540375 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540375 Supplier Invitrogen™ Supplier No. PA540375
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MSSYFVNSFS GRYPNGPDYQ LLNYGSGSSL SGSYRDPAAM HTGSYGYNYN Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

The Hox proteins are a family of transcription factors that play a role in development and cellular differentiation by regulating downstream target genes. Specifically, the Hox proteins direct DNA-protein and protein-protein interactions that assist in determining the morphologic features associated with the anterior-posterior body axis. Hox proteins are involved in controlling axial patterning, leukemias and hereditary malformations. HoxB5 (homeobox protein Hox-B5), also known as HOX2, HU-1, HOX2A, Hox2.1 or HHO.C10, is a member of the Antp homeobox (Hox) family. It is a 269 amino acid long nuclear protein expressed in the central nervous system. HoxB5 contains one homeobox DNAbinding domain and plays a role in the regulation of lung and gut development, providing cells with positional identities on the anterior-posterior body axis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXB5
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HOXB5
Gene Accession No. P09067
Gene Alias AI385717; HHO.C10; homeo box 2A; homeo box B5; homeobox B5; homeobox protein H24.1; Homeobox protein HHO.C10; homeobox protein Hox-2.1; Homeobox protein Hox-2A; homeobox protein Hox-B5; Homeobox protein Hu-1; homeobox protein Mu-1; HOX2; Hox2.1; Hox-2.1; HOX2A; HOXB5; Hoxb-5; HU-1; RGD1562292
Gene Symbols HOXB5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human HOXB5 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3215
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.