Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXB5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599414
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599414 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599414 Supplier Thermo Scientific Supplier No. PA599414
Add to Cart
Add to Cart

HOXB5 Polyclonal Antibody

HOXB5 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXB5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HOXB5
Gene Accession No. P09067, P09079
Gene Alias AI385717; HHO.C10; homeo box 2A; homeo box B5; homeobox B5; homeobox protein H24.1; Homeobox protein HHO.C10; homeobox protein Hox-2.1; Homeobox protein Hox-2A; homeobox protein Hox-B5; Homeobox protein Hu-1; homeobox protein Mu-1; HOX2; Hox2.1; Hox-2.1; HOX2A; HOXB5; Hoxb-5; HU-1; RGD1562292
Gene Symbols HOXB5
Host Species Rabbit
Immunogen A synthesized peptide derived from human HOXB5(Accession P09067), corresponding to amino acid residues T164-H214.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15413, 3215
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.