Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXB5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111718
Description
Immunogen sequence: ASSSHFGAVG ESSRAFPAPA QEPRFRQAAS SCSLSSPESL PCTNGDSHGA KPSASSPSDQ ATSASSSANF TEIDEASASS EPEEAASQLS SPSLARAQPE PMATSTAAPE GQTPQIFPWM R.
The Hox proteins are a family of transcription factors that play a role in development and cellular differentiation by regulating downstream target genes. Specifically, the Hox proteins direct DNA-protein and protein-protein interactions that assist in determining the morphologic features associated with the anterior-posterior body axis. Hox proteins are involved in controlling axial patterning, leukemias and hereditary malformations. HoxB5 (homeobox protein Hox-B5), also known as HOX2, HU-1, HOX2A, Hox2.1 or HHO.C10, is a member of the Antp homeobox (Hox) family. It is a 269 amino acid long nuclear protein expressed in the central nervous system. HoxB5 contains one homeobox DNAbinding domain and plays a role in the regulation of lung and gut development, providing cells with positional identities on the anterior-posterior body axis.
Specifications
HOXB5 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P09067 | |
HOXB5 | |
Recombinant Protein corresonding to Human HOXB5. Recombinant protein control fragment (Product #RP-107998). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry | |
Unconjugated | |
HOXB5 | |
AI385717; HHO.C10; homeo box 2A; homeo box B5; homeobox B5; homeobox protein H24.1; Homeobox protein HHO.C10; homeobox protein Hox-2.1; Homeobox protein Hox-2A; homeobox protein Hox-B5; Homeobox protein Hu-1; homeobox protein Mu-1; HOX2; Hox2.1; Hox-2.1; HOX2A; HOXB5; Hoxb-5; HU-1; RGD1562292 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3215 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction