Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXB6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116164
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116164 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116164 Supplier Invitrogen™ Supplier No. PA5116164
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOXB6 Polyclonal Antibody for IHC (P), Western Blot

HOXB6 belongs to the homeobox family. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXB genes located in a cluster on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXB6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene HOXB6
Gene Accession No. P09023, P17509
Gene Alias homeo box 2B; homeo box B6; homeobox B6; homeobox protein Hox-2.2; Homeobox protein Hox-2B; homeobox protein Hox-B6; Homeobox protein Hu-2; homeobox protein MH-22A; HOX2; Hox-2.2; HOX2B; Hoxb6; Hoxb-6; HU-2; RGD1562142
Gene Symbols HOXB6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 128-155 amino acids from the Central region of human HOXB6.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15414, 3216
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.