Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXB6 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA537867
Description
This antibody is predicted to react with mouse, rat, dog, pig and cow based on sequence homology.
Specifications
| HOXB6 | |
| Polyclonal | |
| Unconjugated | |
| HOXB6 | |
| homeobox B6, HOX2, HOX2B, HU-2, Hox-2.2, homeo box 2B, homeo box B6, homeobox protein Hox-2.2, homeobox protein Hox-2B, homeobox protein Hox-B6, homeobox protein Hu-2 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 3216 | |
| -20°C | |
| Liquid |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P17509 | |
| HOXB6 | |
| Peptide with sequence EPRKSDCAQDKS, from the internal region of HOXB6 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction