Learn More
Invitrogen™ HOXB7 Polyclonal Antibody

Description
HOXB7 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | HOXB7 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Hoxb7 |
| Gene Accession No. | P18864 |
| Gene Alias | AI325018; HHO.C1; homeo box 2C; homeo box B7; homeo box c1 protein; homeobox B7; homeobox gene B7; homeobox protein HHO.C1; Homeobox protein Hox-2.3; homeobox protein Hox-2C; Homeobox protein Hox-B7; homeobox protein MH-22B; Homeobox protein MuB1; homeobox protein R1B; HOX2; Hox-2.3; Hox-23; HOX2C; Hox2r1b; Hoxb7; Hoxb-7; R1b |
| Gene Symbols | Hoxb7 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.