Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXB8 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299554MFV610
Description
HOXB8 Polyclonal antibody specifically detects HOXB8 in Mouse samples. It is validated for Western BlotSpecifications
HOXB8 | |
Polyclonal | |
Western Blot | |
homeo box 2D, homeo box B8, homeobox B8, Homeobox protein Hox-2.4, Homeobox protein Hox-2D, homeobox protein Hox-B8, HOX2, HOX2DHox-2.4 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HOXB8 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
3218 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction