Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXB8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987321
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7321 Supplier Bioss Supplier No. BS6539R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is a member of the Antp homeobox family and encodes a nuclear protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox B genes located on chromosome 17. The encoded protein functions as a sequence-specific transcription factor that is involved in development. Increased expression of this gene is associated with colorectal cancer. Mice that have had the murine ortholog of this gene knocked out exhibit an excessive pathologic grooming behavior. This behavior is similar to the behavior of humans suffering from the obsessive-compulsive spectrum disorder trichotillomania.

Specifications

Antigen HOXB8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HOXB8
Gene Accession No. P17481
Gene Alias homeo box 2D; homeo box B8; homeobox B8; homeobox gene B8; Homeobox protein Hox-2.4; Homeobox protein Hox-2D; Homeobox protein Hox-B8; homeobox protein R1a; HOX2; Hox-2.4; Hox2.4 homeobox homolog; HOX2D; Hox2r1a; Hoxb8; Hoxb-8
Gene Symbols HOXB8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HOXB8 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3218
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.