Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXB8B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572625
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572625 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572625 Supplier Invitrogen™ Supplier No. PA572625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOXB8B Polyclonal Antibody for Western Blot

Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXB8B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene hoxb8b
Gene Accession No. Q8JH55
Gene Alias homeo box B8b; homeobox b8b; homeobox gene A-8; homeobox gene B-8b; homeobox protein Hox-A8; Homeobox protein Hox-B8b; hoxa8; hox-A8; Hoxa8 protein; hoxb8b; z-9
Gene Symbols hoxb8b
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 186-212 amino acids from the C-terminal region of DANRE hoxb8b.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30420
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.