Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXB9 Antibody (SR1469), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | HOXB9 |
---|---|
Clone | SR1469 |
Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
HOXB9 Monoclonal antibody specifically detects HOXB9 in Human samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
HOXB9 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3219 | |
IgG | |
Affinity purified |
SR1469 | |
Western Blot, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
homeo box 2E, homeo box B9, homeobox B9, Homeobox protein Hox-2.5, Homeobox protein Hox-2E, homeobox protein Hox-B9, HOX-2.5, HOX2EHOX2 | |
A synthesized peptide derived from human HOXB9 (Uniprot #: P17482) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title