Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXB9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540576
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540576 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540576 Supplier Invitrogen™ Supplier No. PA540576
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SPHASGSLPS VYHPYIQPQG VPPAESRYLR TWLEPAPRGE AAPGQGQAAV Sequence homology: Cow: 100%; Dog: 93%; Horse: 100%; Human: 100%; Mouse: 79%; Pig: 100%; Rabbit: 100%; Rat: 79%.

HOXB9 belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. HOXB9 is one of several homeobox HOXB genes located in a cluster on chromosome 17. The exact role of this gene has yet to be determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXB9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HOXB9
Gene Accession No. P17482
Gene Alias HGNC:5120; homeo box 2E; homeo box B9; homeobox B9; homeobox protein Hox-2.5; Homeobox protein Hox-2E; homeobox protein Hox-B9; HOX2; HOX-2.5; HOX2E; HOXB9; Hoxb-9; MGC124051; MGC124052; MGC124053; MGC124054
Gene Symbols HOXB9
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human HOXB9 (aa 67-116).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3219
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.