Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXC10 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171933AF532
Description
HOXC10 Polyclonal antibody specifically detects HOXC10 in Human samples. It is validated for Western BlotSpecifications
HOXC10 | |
Polyclonal | |
Western Blot | |
homeo box C10, homeobox C10, Homeobox protein Hox-3I, homeobox protein Hox-C10, homeoprotein C10, HOX3I, MGC5259 | |
The immunogen for this product maps to a region between residue 100 and 150 of human Homeobox C10 using the numbering given in entry NP_059105.2 (GeneID 3226). | |
0.1 mL | |
Cell Cycle and Replication | |
3226 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction