Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXC10 Antibody (OTI5C10), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270909C
Description
HOXC10 Monoclonal specifically detects HOXC10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
HOXC10 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HOXC10 | |
Recombinant protein fragment corresponding to amino acids 1-262 of human HOXC10 (NP_059105) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI5C10 | |
Western Blot, Immunohistochemistry | |
homeo box C10, homeobox C10, Homeobox protein Hox-3I, homeobox protein Hox-C10, homeoprotein C10, HOX3I, MGC5259 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
3226.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction