Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXC10 Monoclonal Antibody (OTI5C10)
GREENER_CHOICE

Catalog No. PIMA527065
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527065 Supplier Invitrogen™ Supplier No. MA527065
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HOXC10 Monoclonal Antibody for Western Blot, IHC (P)

This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The protein level is controlled during cell differentiation and proliferation, which may indicate this protein has a role in origin activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXC10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5C10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HOXC10
Gene Accession No. Q9NYD6
Gene Alias AI506621; homeo box C10; homeobox C10; homeobox protein Hox-3.6; Homeobox protein Hox-3I; homeobox protein Hox-C10; homeoprotein C10; Hox3.5 homeobox; Hox-3.6; HOX3I; HOXC10; Hoxc-10
Gene Symbols HOXC10
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-262 of HOXC10 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3226
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.