Learn More
HOXC10 Mouse anti-Human, Clone: OTI1H5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808866

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The protein level is controlled during cell differentiation and proliferation, which may indicate this protein has a role in origin activation.Specifications
HOXC10 | |
Monoclonal | |
Unconjugated | |
HOXC10 | |
HOX3I | |
Mouse | |
Affinity Chromatography | |
RUO | |
3226 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1H5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NYD6 | |
HOXC10 | |
Human recombit protein fragment corresponding to amino acids 1-262 of human HOXC10 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.