Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXC10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584176
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584176 Supplier Invitrogen™ Supplier No. PA584176
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTCPRNVTPN SYAEPLAAPG GGERYSRSAG MYMQSGSDFN CGVMRGCGLA PSLSKRDEGS SPSLALNTYP SYLSQLDSWG DPKAAYRLEQ P Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 96%.

This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The protein level is controlled during cell differentiation and proliferation, which may indicate this protein has a role in origin activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXC10
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HOXC10
Gene Accession No. Q9NYD6
Gene Alias AI506621; homeo box C10; homeobox C10; homeobox protein Hox-3.6; Homeobox protein Hox-3I; homeobox protein Hox-C10; homeoprotein C10; Hox3.5 homeobox; Hox-3.6; HOX3I; HOXC10; Hoxc-10
Gene Symbols HOXC10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HOXC10. Recombinant protein control fragment (Product #RP-101521).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3226
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.