Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXC11 Monoclonal Antibody (OTI2A10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502574S


Description
HOXC11 Monoclonal antibody specifically detects HOXC11 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
HOXC11 | |
Monoclonal | |
0.71 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O43248 | |
Mouse | |
Affinity chromatography | |
RUO | |
3227 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI2A10 | |
Unconjugated | |
HOXC11 | |
homeo box C11; homeobox C11; homeobox protein Hox-3.7; Homeobox protein Hox-3H; homeobox protein Hox-C11; homeobox protein Hox-C11-like; Hox-3.7; HOX3H; HOXC11; Hoxc-11; LOC100911859; LOW QUALITY PROTEIN: homeobox protein Hox-C11; MGC4906 | |
Full length human recombinant protein of human HOXC11 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction