Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXC11 Monoclonal Antibody (OTI3E10)
GREENER_CHOICE

Catalog No. PIMA525460
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525460 Supplier Invitrogen™ Supplier No. MA525460
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HOXC11 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The product of this gene binds to a promoter element of the lactase-phlorizin hydrolase. It also may play a role in early intestinal development. An alternatively spliced variant encoding a shorter isoform has been described but its full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXC11
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3E10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HOXC11
Gene Accession No. O43248
Gene Alias homeo box C11; homeobox C11; homeobox protein Hox-3.7; Homeobox protein Hox-3H; homeobox protein Hox-C11; homeobox protein Hox-C11-like; Hox-3.7; HOX3H; HOXC11; Hoxc-11; LOC100911859; LOW QUALITY PROTEIN: homeobox protein Hox-C11; MGC4906
Gene Symbols HOXC11
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-304 of HOXC11 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3227
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.