Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    HOXC11 Rabbit anti-Human, Polyclonal, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304552100UL
Description
HOXC11 Polyclonal specifically detects HOXC11 in Human samples. It is validated for Western Blot.Specifications
| HOXC11 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 3227 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG | 
| Western Blot | |
| Unconjugated | |
| homeo box C11, homeobox C11, Homeobox protein Hox-3H, homeobox protein Hox-C11, HOX3H, MGC4906 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human HOXC11 (NP_055027.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            