Learn More
Invitrogen™ HOXC12 Monoclonal Antibody (3E1)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA519125
Catalog No. PIMA519125
Only null left
Add to Cart
Description
Description
Peptide Sequence: MGEHNLLNPG FVGPLVNIHT GDTFYFPNFR ASGAQLPGLP SLSYPRRDNV CSLSWPSAEP CNGYPQPYLG SPVSLNPPFG RTCELARVED GKGYYREPCA.
The Homeobox (Hox) proteins are a family of transcription factors that play a role in development and cellular differentiation by regulating downstream target genes. Specifically, the Hox proteins direct DNA-protein and protein-protein interactions that assist in determining the morphologic features associated with the anterior-posterior body axis. Hox proteins are involved in controlling axial patterning, leukemias and hereditary malformations. In mammals, there are four gene clusters encoding Hox proteins, namely HoxA, HoxB, HoxC and HoxD, that are located on different chromosomes, each consisting of 9-11 tandemly arranged genes. HoxC12 (Homeobox C12), also known as Hox3, Hox3F or HOC3F, is a member of the Abd-B homeobox (Hox) family and is encoded by a gene from the HoxC cluster. HoxC12 is a 282 amino acid long nuclear protein that contains one homeobox DNA-binding domain.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HOXC12
Monoclonal
0.5 mg/mL
PBS with no preservative; pH 7.4
P31275
HOXC12
HOXC12 (NP_776272, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Western Blot, Immunocytochemistry
3E1
Unconjugated
HOXC12
HOC3F; homeo box C12; homeobox C12; homeobox protein Hox-3.8; Homeobox protein Hox-3F; Homeobox protein Hox-C12; Homeobox protein Hox-C12 (Hox-3F); HOX3; Hox-3.8; HOX3F; HOXC12; Hoxc-12
Mouse
Affinity chromatography
RUO
3228
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.