Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXC13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562447
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562447 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562447 Supplier Invitrogen™ Supplier No. PA562447
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSFASSYQAM PGYLDVSVVP GISGHPEPRH DALIPVEGYQ HWALSNGWDS QVYCSKE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The product of this gene may play a role in the development of hair, nail, and filiform papilla.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXC13
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HOXC13
Gene Accession No. P31276
Gene Alias ECTD9; homeo box 3G; homeo box C13; homeobox C13; homeobox protein Hox-3G; Homeobox protein Hox-C13; HOX3; HOX3G; HOXC13; Hoxc-13; NUP98/HOXC13; RGD1563984
Gene Symbols HOXC13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HOXC13. Recombinant protein control fragment (Product #RP-95630).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3229
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.