Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXC4 Monoclonal Antibody (OTI1F6)
GREENER_CHOICE

Catalog No. PIMA527195
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527195 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527195 Supplier Invitrogen™ Supplier No. MA527195
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HOXC4 Monoclonal Antibody for Western Blot, IHC (P)

Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXC4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HOXC4
Gene Accession No. P09017
Gene Alias cp19; homeo box 3E; homeo box C4; homeobox C4; Homeobox gene C4; homeobox protein CP19; Homeobox protein Hox-3.5; homeobox protein Hox-3E; Homeobox protein Hox-C4; homeobox protein R3; HOX3; Hox-3.5; Hox-35; HOX3E; Hox3r3; Hoxc4; Hoxc-4
Gene Symbols HOXC4
Host Species Mouse
Immunogen Full length human recombinant protein of HOXC4 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3221
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.