Learn More
HOXC4 Mouse anti-Human, Clone: OTI6B11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809706

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.Specifications
HOXC4 | |
Monoclonal | |
Unconjugated | |
HOXC4 | |
HOX3, HOX3E, cp19 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3221 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI6B11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P09017 | |
HOXC4 | |
Full length human recombit protein of human HOXC4 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.