Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXC4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589486
Description
Immunogen sequence: PEHSPEYYGR TRESGFQHHH QELYPPPPPR PSYPERQYSC TSLQGPGNSR GHGPAQAGHH HPEKSQSLCE PAPLSGASAS PSPAPPACSQ PAPDHPSSAA S; Positive Samples: Mouse liver, Rat liver; Cellular Location: Nucleus.
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
Specifications
HOXC4 | |
Polyclonal | |
Unconjugated | |
HOXC4 | |
cp19; homeo box 3E; homeo box C4; homeobox C4; Homeobox gene C4; homeobox protein CP19; Homeobox protein Hox-3.5; homeobox protein Hox-3E; Homeobox protein Hox-C4; homeobox protein R3; HOX3; Hox-3.5; Hox-35; HOX3E; Hox3r3; Hoxc4; Hoxc-4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
15423, 24459 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P18865, Q08624 | |
HOXC4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 30-130 of human HOXC4 (NP_0554352). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction