Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXC4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572627
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572627 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572627 Supplier Invitrogen™ Supplier No. PA572627
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOXC4A Polyclonal Antibody for Western Blot

Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXC4A
Applications Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene hoxc4a
Gene Accession No. Q9PWM3
Gene Alias homeo box C4a; homeobox c4a; homeobox gene C-4; homeobox protein Hox-C4a; hoxc4; Hox-C4; Hoxc4 protein; hoxc4a; si:dkey-81p22.8; z-96; zgc:110513
Gene Symbols hoxc4a
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 240-267 amino acids from the C-terminal region of DANRE hoxc4a.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30345
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.