Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXC5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981496
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1496 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1496 Supplier Bioss Supplier No. BS11589R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.

Specifications

Antigen HOXC5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HOXC5
Gene Accession No. Q00444
Gene Alias CP11; homeo box C5; homeobox C5; Homeobox protein CP11; Homeobox protein Hox-3D; Homeobox protein Hox-C5; HOX3; Hox-34; HOX3D; HOXC5
Gene Symbols HOXC5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HOXC5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3222
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.