Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541479
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541479 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541479 Supplier Invitrogen™ Supplier No. PA541479
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KIWFQNRRMK WKKESNLTST LSGGGGGATA DSLGGKEEKR EETEEEKQKE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Sheep: 100%.

This protein belongs to the homeobox family, members of which encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene, HOXC6, is one of several HOXC genes located in a cluster on chromosome 12. Three genes, HOXC5, HOXC4 and HOXC6, share a 5' non-coding exon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXC6
Applications Immunohistochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HOXC6
Gene Accession No. P10629
Gene Alias Antennapedia-type homeotic factor; CP25; HHO.C8; homeo box 3C; homeo box C6; homeo box C8 protein; homeobox C6; homeobox protein CP25; Homeobox protein HHO.C8; homeobox protein Hox-3.3; Homeobox protein Hox-3C; homeobox protein Hox-6.1; Homeobox protein Hox-C6; Hox C6; HOX3; Hox-3.3; HOX3C; Hox-6.1; Hoxc6; Hoxc-6
Gene Symbols HOXC6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human HOXC6 (aa 186-235).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15425
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.